accessibility menu, dialog, popup

UserName

H3CPO

CAS RN 707-61-9

IUPAC Name: 4-methyl-1-phenyl-2,3-dihydro-1H-1λ⁵-phosphol-1-one
Synonyms: 3-methyl-1-phenyl-4,5-dihydro-1λ⁵-phosphol-1-one
Molecular Weight (g/mol): 192.2
Molecular Formula: C11H13OP
InChi Key: YMKWWHFRGALXLE-UHFFFAOYNA-N
SMILES: CC1=CP(=O)(CC1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)