accessibility menu, dialog, popup

UserName

CAS RN 705-86-2

IUPAC Name: 6-pentyloxan-2-one
Synonyms: delta-decalactone
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: GHBSPIPJMLAMEP-UHFFFAOYNA-N
SMILES: CCCCCC1CCCC(=O)O1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)