missing translation for 'onlineSavingsMsg'
Learn More

delta-Decanolactone 97.0+%, TCI America™

Catalog No. D141625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D141625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D141625ML Supplier TCI America Supplier No. D141625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 705-86-2
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00006649
InChI Key GHBSPIPJMLAMEP-UHFFFAOYNA-N
Synonym delta-decalactone, 6-pentyltetrahydro-2h-pyran-2-one, 5-decanolide, decanolide-1,5, 5-decalactone, decan-5-olide, amyl-delta-valerolactone, delta-decanolactone, 5-pentyl-5-pentanolide, 2h-pyran-2-one, tetrahydro-6-pentyl
PubChem CID 12810
ChEBI CHEBI:87327
IUPAC Name 6-pentyloxan-2-one
SMILES CCCCCC1CCCC(=O)O1

Specifications

Color Yellow
Boiling Point 123°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material delta-Decanolactone
EINECSNumber (5)-0698
RTECSNumber UQ1355000
TSCA Yes