missing translation for 'onlineSavingsMsg'
Learn More

delta-Decanolactone, 97%

Catalog No. AAA1175636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1175636 500 g
AAA1175614 25 g
AAA1175622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1175636 Supplier Thermo Scientific Chemicals Supplier No. A1175636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 705-86-2 | C10H18O2 | 170.25 g/mol

Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

Solubility
Soluble in water (4 mg/ml at 28°C), alcohols, and propylene glycol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 705-86-2
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00006649
InChI Key GHBSPIPJMLAMEP-UHFFFAOYNA-N
Synonym delta-decalactone, 6-pentyltetrahydro-2h-pyran-2-one, 5-decanolide, decanolide-1,5, 5-decalactone, decan-5-olide, amyl-delta-valerolactone, delta-decanolactone, 5-pentyl-5-pentanolide, 2h-pyran-2-one, tetrahydro-6-pentyl
PubChem CID 12810
ChEBI CHEBI:87327
IUPAC Name 6-pentyloxan-2-one
SMILES CCCCCC1CCCC(=O)O1

Specifications

Melting Point -25°C
Density 0.971
Boiling Point 117°C to 120°C (0.02 mmHg)
Flash Point 145°C (293°F)
Refractive Index 1.458
Quantity 500 g
Beilstein 117520
Solubility Information Soluble in water (4mg/ml at 28°C),alcohols,and propylene glycol.
Formula Weight 170.25
Percent Purity 97%
Chemical Name or Material delta-Decanolactone
Show More Show Less
EINECSNumber 211-889-1
RTECSNumber UQ1355000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only