accessibility menu, dialog, popup

UserName

CAS RN 70361-61-4

H3COOSHNH2HCl

CAS RN 70361-61-4

IUPAC Name: methyl 2-amino-3-sulfanylpropanoate hydrochloride
Synonyms: methyl 2-amino-3-sulfanylpropanoate hydrochloride
Molecular Weight (g/mol): 171.64
Molecular Formula: C4H10ClNO2S
InChi Key: WHOHXJZQBJXAKL-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CS
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results