missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Cysteine Methyl Ester Hydrochloride 98.0+%, TCI America™
$283.52 - $694.54
Chemical Identifiers
| CAS | 70361-61-4 |
|---|---|
| Molecular Formula | C4H10ClNO2S |
| Molecular Weight (g/mol) | 171.639 |
| MDL Number | MFCD00173915 |
| InChI Key | WHOHXJZQBJXAKL-AENDTGMFSA-N |
| Synonym | H-D-Cys-OMe.HCl |
| PubChem CID | 637545 |
| IUPAC Name | methyl (2S)-2-amino-3-sulfanylpropanoate;hydrochloride |
| SMILES | COC(=O)C(CS)N.Cl |
Chemical Identifiers
| 70361-61-4 | |
| 171.639 | |
| WHOHXJZQBJXAKL-AENDTGMFSA-N | |
| 637545 | |
| COC(=O)C(CS)N.Cl |
| C4H10ClNO2S | |
| MFCD00173915 | |
| H-D-Cys-OMe.HCl | |
| methyl (2S)-2-amino-3-sulfanylpropanoate;hydrochloride |
Specifications
| 70361-61-4 | |
| White-Yellow | |
| MFCD00173915 | |
| H-D-Cys-OMe.HCl | |
| COC(=O)C(CS)N.Cl | |
| 171.639 | |
| 171.64 | |
| Crystalline Powder |
| 135°C | |
| C4H10ClNO2S | |
| 5 g | |
| WHOHXJZQBJXAKL-AENDTGMFSA-N | |
| methyl (2S)-2-amino-3-sulfanylpropanoate;hydrochloride | |
| 637545 | |
| ≥98.0% (T) | |
| D-Cysteine Methyl Ester Hydrochloride |
Safety and Handling
TSCA : No