accessibility menu, dialog, popup

UserName

OH(R)(R)OHHOOOHOBrNHNNHNN

CAS RN 70359-46-5

IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid; 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
Synonyms: L(+)-tartaric acid; brimonidine
Molecular Weight (g/mol): 442.23
Molecular Formula: C15H16BrN5O6
InChi Key: QZHBYNSSDLTCRG-LREBCSMRSA-N
SMILES: O[C@H]([C@@H](O)C(O)=O)C(O)=O.BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results