accessibility menu, dialog, popup

UserName

CH3H3COBr

CAS RN 701-07-5

IUPAC Name: 1-bromo-2-(propan-2-yloxy)benzene
Synonyms: 1-bromo-2-isopropoxybenzene
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: MMORVPBHAHXAHH-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=CC=C1Br
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)