accessibility menu, dialog, popup

UserName

HO

CAS RN 700-57-2

IUPAC Name: adamantan-2-ol
Synonyms: adamantan-2-ol
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: FOWDOWQYRZXQDP-UHFFFAOYSA-N
SMILES: OC1C2CC3CC(C2)CC1C3
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)