missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanol 98.0+%, TCI America™

Catalog No. A07205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07205G 5 g
A072025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07205G Supplier TCI America Supplier No. A07205G

Chemical Identifiers

CAS 700-57-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00074744
InChI Key FOWDOWQYRZXQDP-UHFFFAOYSA-N
Synonym 2-adamantanol, 2-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-2-ol, tricyclo 3.3.1.1∼3,7∼ decan-2-ol, 1r,3r,5r,7r-adamantan-2-ol, 1-adamantanol;, 2alpha-adamantanol, pubchem8753
PubChem CID 64149
IUPAC Name adamantan-2-ol
SMILES C1C2CC3CC1CC(C2)C3O

Specifications

Color White
Quantity 5 g
Formula Weight 152.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Adamantanol