missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanol, 98%

Catalog No. AC167310050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167310050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC167310050 Supplier Thermo Scientific Chemicals Supplier No. 167310050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 700-57-2 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 700-57-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00074744
InChI Key FOWDOWQYRZXQDP-UHFFFAOYSA-N
Synonym 2-adamantanol, 2-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-2-ol, tricyclo 3.3.1.1∼3,7∼ decan-2-ol, 1r,3r,5r,7r-adamantan-2-ol, 1-adamantanol;, 2alpha-adamantanol, pubchem8753
PubChem CID 64149
IUPAC Name adamantan-2-ol
SMILES C1C2CC3CC1CC(C2)C3O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 258.0°C to 265.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol, diethyl ether, methylene, chloride and acetone
Formula Weight 152.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Adamantanol, 98%
Show More Show Less
EINECSNumber 211-846-7

RUO – Research Use Only