accessibility menu, dialog, popup

UserName

NH2NFNNHN

CAS RN 700-49-2

IUPAC Name: 2-fluoro-7H-purin-6-amine
Synonyms: 2-fluoroadenine
Molecular Weight (g/mol): 153.12
Molecular Formula: C5H4FN5
InChi Key: WKMPTBDYDNUJLF-UHFFFAOYSA-N
SMILES: NC1=C2NC=NC2=NC(F)=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)