missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-L-tyrosine, 98%, Spectrum™ Chemical

Catalog No. 18600129
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600129 5 g Amber Glass Bottle
18600128 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600129 Supplier Spectrum Chemical Mfg Cor Supplier No. I30385GM
Get Up to $500 in Chemicals with Purchase
View all promotions

I3038, 70-78-0, C9H10INO3

Spectrum™ Chemical 3-Iodo-L-tyrosine

Chemical Identifiers

CAS 70-78-0
Molecular Formula C9H10INO3
Molecular Weight (g/mol) 307.09
MDL Number MFCD00002608
InChI Key UQTZMGFTRHFAAM-JLDDOWRYNA-N
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98%
Grade Reagent