missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Iodo-L-tyrosine, 97%

Catalog No. AC122440250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122440250 25 g Glass Bottle
AC122440010 1 g Glass Bottle
AC122440050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC122440250 Supplier Thermo Scientific Chemicals Supplier No. 122440250

CAS: 70-78-0 | C9H10INO3 | 307.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70-78-0
Molecular Formula C9H10INO3
Molecular Weight (g/mol) 307.09
MDL Number MFCD00002608
InChI Key UQTZMGFTRHFAAM-JLDDOWRYNA-N
Synonym 3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
PubChem CID 439744
ChEBI CHEBI:27847
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O

Specifications

Melting Point 196.0°C to 198.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Specific Rotation − 3.18
Merck Index 15, 5090
Solubility Information Solubility in water: .. Other solubilities: soluble in 15 parts boiling water
Water 1% max. (K.F.)
Quantity 25 g
Formula Weight 307.08
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Iodo-L-tyrosine
Show More Show Less

RUO – Research Use Only