accessibility menu, dialog, popup

UserName

CH3OOH

CAS RN 70-70-2

IUPAC Name: 1-(4-hydroxyphenyl)propan-1-one
Synonyms: mepal frenohypon sterofenon paraoxypropiophenone frenon POP paroxipropiona bio-fren possipione proxiphenon
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: RARSHUDCJQSEFJ-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)