missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxypropiophenone, 99%

Catalog No. AAA1531436
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1531436 500 g
AAA1531422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1531436 Supplier Thermo Scientific Chemicals Supplier No. A1531436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70-70-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70-70-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002361
InChI Key RARSHUDCJQSEFJ-UHFFFAOYSA-N
Synonym 4'-hydroxypropiophenone, paroxypropione, 4-hydroxypropiophenone, 1-4-hydroxyphenyl propan-1-one, mepal, paroxypropion, frenantol, frenohypon, p-hydroxypropiophenone, proxiphenon
PubChem CID 6271
ChEBI CHEBI:32352
IUPAC Name 1-(4-hydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 148°C to 151°C
Flash Point 180°C (356°F)
Quantity 500 g
Beilstein 907511
Merck Index 14,7044
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 4'-Hydroxypropiophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-743-2
RTECSNumber UH1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only