accessibility menu, dialog, popup

UserName

CAS RN 6999-19-5

CH3SiH3CCH3CH3OH

CAS RN 6999-19-5

IUPAC Name: 4-(trimethylsilyl)but-3-yn-2-ol
Synonyms: 4-(trimethylsilyl)but-3-yn-2-ol
Molecular Weight (g/mol): 142.27
Molecular Formula: C7H14OSi
InChi Key: HJJSDJHRTMFJLP-UHFFFAOYNA-N
SMILES: CC(O)C#C[Si](C)(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results