missing translation for 'onlineSavingsMsg'
Learn More

4-Trimethylsilyl-3-butyn-2-ol 97.0+%, TCI America™

Catalog No. T19265G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19265G Supplier TCI America Supplier No. T19265G

Chemical Identifiers

CAS 6999-19-5
Molecular Formula C7H14OSi
Molecular Weight (g/mol) 142.273
MDL Number MFCD00190213
InChI Key HJJSDJHRTMFJLP-UHFFFAOYSA-N
Synonym 4-trimethylsilyl-3-butyn-2-ol, 4-trimethylsilyl but-3-yn-2-ol, 1-trimethylsilylbut-1-yne-3-ol, +/-4-trimethylsilyl-3-butyn-2-ol, acmc-20mpjc, 3-butyn-2-ol, 4-trimethylsilyl-, 2s, acmc-1bfey, hjjsdjhrtmfjlp-uhfffaoysa, 3-butyn-2-ol,4-trimethylsilyl
PubChem CID 2760828
IUPAC Name 4-trimethylsilylbut-3-yn-2-ol
SMILES CC(C#C[Si](C)(C)C)O

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 142.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Trimethylsilyl-3-butyn-2-ol
TSCA No
Recommended Storage Refrigerator