missing translation for 'onlineSavingsMsg'
Learn More

4-Trimethylsilyl-3-butyn-2-ol, 97%

Catalog No. AAL1481514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1481514 25 g
AAL1481506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1481514 Supplier Thermo Scientific Chemicals Supplier No. L1481514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6999-19-5 | C7H14OSi | 142.273 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6999-19-5
Molecular Formula C7H14OSi
Molecular Weight (g/mol) 142.273
MDL Number MFCD00190213
InChI Key HJJSDJHRTMFJLP-UHFFFAOYSA-N
Synonym 4-trimethylsilyl-3-butyn-2-ol, 4-trimethylsilyl but-3-yn-2-ol, 1-trimethylsilylbut-1-yne-3-ol, +/-4-trimethylsilyl-3-butyn-2-ol, acmc-20mpjc, 3-butyn-2-ol, 4-trimethylsilyl-, 2s, acmc-1bfey, hjjsdjhrtmfjlp-uhfffaoysa, 3-butyn-2-ol,4-trimethylsilyl
PubChem CID 2760828
IUPAC Name 4-trimethylsilylbut-3-yn-2-ol
SMILES CC(C#C[Si](C)(C)C)O

Specifications

Boiling Point 126°C to 129°C
Quantity 25 g
UN Number UN1987
Beilstein 1923632
Flash Point 57°C (135°F)
Formula Weight 142.27
Percent Purity 97%
Refractive Index 1.446
Density 0.846
Chemical Name or Material 4-Trimethylsilyl-3-butyn-2-ol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only