accessibility menu, dialog, popup

UserName

CH3(S)(R)OH(S)H3C(S)(S)H3C(S)(R)OH(R)(S)(S)CH3OH3COCH3H3CHOO(Z)

CAS RN 6990-06-3

IUPAC Name: 2-[(1Z,2S,3aS,3bS,5aS,6S,7R,9aS,9bS,10R,11aR)-2-(acetyloxy)-7,10-dihydroxy-3a,3b,6,9a-tetramethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-ylidene]-6-methylhept-5-enoic acid
Synonyms: fusidic acid discoid furoβ frumil fucidin fluss rosis desal (-)-fusidic acid urian
Molecular Weight (g/mol): 516.72
Molecular Formula: C31H48O6
InChi Key: IECPWNUMDGFDKC-MZJAQBGESA-N
SMILES: C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]1(C)[C@H]2[C@H](O)C[C@H]2\C([C@H](C[C@]12C)OC(C)=O)=C(/CCC=C(C)C)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)