missing translation for 'onlineSavingsMsg'
Learn More

Fusidic acid, 98%, Thermo Scientific Chemicals

Catalog No. AC458460050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458460050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458460050 Supplier Thermo Scientific Chemicals Supplier No. 458460050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6990-06-3

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C31H48O6
Quantity 5 g
Synonym methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform
InChI Key DIWVBIXQCNRCFE-UHFFFAOYNA-N
SMILES COC(C1=CC=CC=C1)C(=O)O
IUPAC Name 2-methoxy-2-phenylacetic acid
Molecular Weight (g/mol) 516.71
PubChem CID 107202
Formula Weight 516.71
Percent Purity 98%
Chemical Name or Material Fusidic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 230-256-

RUO – Research Use Only