accessibility menu, dialog, popup

UserName

CH3NONHOH2NH2N

CAS RN 6972-82-3

IUPAC Name: 5,6-diamino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5,6-diamino-1-methyl-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 156.15
Molecular Formula: C5H8N4O2
InChi Key: PSIJQVXIJHUQPJ-UHFFFAOYSA-N
SMILES: CN1C(=O)NC(=O)C(N)=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results