accessibility menu, dialog, popup

UserName

OHOO

CAS RN 6972-79-8

IUPAC Name: 1,3-bis(benzyloxy)propan-2-ol
Synonyms: 1,3-bis(benzyloxy)propan-2-ol
Molecular Weight (g/mol): 272.34
Molecular Formula: C17H20O3
InChi Key: ARLSYSVVBAMYKA-UHFFFAOYSA-N
SMILES: OC(COCC1=CC=CC=C1)COCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results