accessibility menu, dialog, popup

UserName

CAS RN 695-12-5

IUPAC Name: ethenylcyclohexane
Synonyms: cyclohexane, ethenyl-
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: LDLDYFCCDKENPD-UHFFFAOYSA-N
SMILES: C=CC1CCCCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)