missing translation for 'onlineSavingsMsg'
Learn More

Vinylcyclohexane 98.0+%, TCI America™

Catalog No. V00225ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00225ML 5 mL
V002225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00225ML Supplier TCI America Supplier No. V00225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 695-12-5
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00001514
InChI Key LDLDYFCCDKENPD-UHFFFAOYSA-N
Synonym vinylcyclohexane, cyclohexylethylene, cyclohexane, ethenyl, cyclohexane, vinyl, cyclohexylethene, unii-q49pj3tt00, cyclohexane, ethenyl-9ci, vinyl cyclohexane, vinyl-cyclohexane, 1-vinylcyclohexane
PubChem CID 12757
IUPAC Name ethenylcyclohexane
SMILES C=CC1CCCCC1

Specifications

Color Colorless
Boiling Point 128°C
Quantity 5 mL
UN Number 3295
Formula Weight 110.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinylcyclohexane
RTECSNumber GV7800000
TSCA No