missing translation for 'onlineSavingsMsg'
Learn More

Vinylcyclohexane, 97%

Catalog No. AAL1075614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1075614 25 g
AAL1075606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1075614 Supplier Thermo Scientific Chemicals Supplier No. L1075614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 695-12-5 | C8H14 | 110.20 g/mol

The use of vinylcyclohexane dioxide (VCD) as a polar dehydrant with subsequent embedment in Spurr was studied. Vinylcyclohexane (VCH) was copolymerized with ethene and propene using methylaluminoxane-activated metallocene catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The use of vinylcyclohexane dioxide (VCD) as a polar dehydrant with subsequent embedment in Spurr was studied. Vinylcyclohexane (VCH) was copolymerized with ethene and propene using methylaluminoxane-activated metallocene catalysts.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 695-12-5
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00001514
InChI Key LDLDYFCCDKENPD-UHFFFAOYSA-N
Synonym vinylcyclohexane, cyclohexylethylene, cyclohexane, ethenyl, cyclohexane, vinyl, cyclohexylethene, unii-q49pj3tt00, cyclohexane, ethenyl-9ci, vinyl cyclohexane, vinyl-cyclohexane, 1-vinylcyclohexane
PubChem CID 12757
IUPAC Name ethenylcyclohexane
SMILES C=CC1CCCCC1

Specifications

Density 0.805
Boiling Point 127°C to 128°C
Flash Point 22°C (71°F)
Refractive Index 1.4465
Quantity 25 g
UN Number UN3295
Beilstein 1304663
Solubility Information Insoluble in water.
Formula Weight 110.2
Percent Purity 97%
Chemical Name or Material Vinylcyclohexane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 211-779-3
RTECSNumber GV7800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only