accessibility menu, dialog, popup

UserName

CAS RN 6940-76-7

IUPAC Name: 1-chloro-3-iodopropane
Synonyms: propane, 1-chloro-3-iodo-
Molecular Weight (g/mol): 204.44
Molecular Formula: C3H6ClI
InChi Key: SFOYQZYQTQDRIY-UHFFFAOYSA-N
SMILES: ClCCCI
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)