accessibility menu, dialog, popup

UserName

CAS RN 68938-72-7

ClClH3CH3CNHNHNNCH3OHOOHOCH3H3CH3CHH

CAS RN 68938-72-7

IUPAC Name: dihydrogen 3-[20-(2-carboxyethyl)-9,14-diethyl-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(21),2,4,6(24),7,9,11,13,15,17,19-undecaen-4-yl]propanoic acid dichloride
Synonyms: dihydrogen 3-[20-(2-carboxyethyl)-9,14-diethyl-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(21),2,4,6(24),7,9,11,13,15,17,19-undecaen-4-yl]propanoic acid dichloride
Molecular Weight (g/mol): 639.62
Molecular Formula: C34H40Cl2N4O4
InChi Key: YUIUEEROGVTICN-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CCC1=C(C)C2=CC3=C(CC)C(C)=C(N3)C=C3N=C(C=C4N=C(C=C1N2)C(C)=C4CCC(O)=O)C(CCC(O)=O)=C3C
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)