accessibility menu, dialog, popup

UserName

CAS RN 68401-81-0

IUPAC Name: (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: ceftizoxime|cefizox|ceftizoxima|ceftix|sodium, ceftizoxime|epocelin|eposerin|ceftizoximum|cefizox in plastic container|ceftizoxime (product)
Molecular Weight (g/mol): 383.4
Molecular Formula: C13H13N5O5S2
InChi Key: NNULBSISHYWZJU-LLKWHZGFSA-N
SMILES: [H][C@]12SCC=C(N1C(=O)[C@H]2NC(=O)C(=N/OC)\C1=CSC(N)=N1)C(O)=O
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)