missing translation for 'onlineSavingsMsg'
Learn More

Ceftizoxime 98.0+%, TCI America™

Catalog No. C26225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26225G 5 g
C26221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26225G Supplier TCI America Supplier No. C26225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68401-81-0
Molecular Formula C13H13N5O5S2
Molecular Weight (g/mol) 383.40
MDL Number MFCD00072000
InChI Key NNULBSISHYWZJU-LLKWHZGFSA-N
PubChem CID 12775087
IUPAC Name (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES [H][C@]12SCC=C(N1C(=O)[C@H]2NC(=O)C(=N/OC)\C1=CSC(N)=N1)C(O)=O

Specifications

Melting Point 227°C
Color White
Quantity 5 g
Formula Weight 383.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ceftizoxime
RTECSNumber XI0367375
TSCA No