accessibility menu, dialog, popup

UserName

BrHNBr

CAS RN 6825-20-3

IUPAC Name: 3,6-dibromo-9H-carbazole
Synonyms: 3,6-dibromo-9H-carbazole
Molecular Weight (g/mol): 325
Molecular Formula: C12H7Br2N
InChi Key: FIHILUSWISKVSR-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NC3=C2C=C(Br)C=C3)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)