missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromocarbazole, 99%

Catalog No. AAH5636014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5636014 25 g
AAH5636006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5636014 Supplier Thermo Scientific Chemicals Supplier No. H5636014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6825-20-3 | C12H7Br2N | 325.003 g/mol

3,6-Dibromocarbazole has been used in the preparation of N-(2-hydroxyethyl)-3,6-dibromocarbazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Dibromocarbazole has been used in the preparation of N-(2-hydroxyethyl)-3,6-dibromocarbazole.

Solubility
Soluble in benzene, chloroform

Notes
Incompatible with oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 6825-20-3
Molecular Formula C12H7Br2N
Molecular Weight (g/mol) 325.003
MDL Number MFCD00004961
InChI Key FIHILUSWISKVSR-UHFFFAOYSA-N
Synonym 3,6-dibromocarbazole, 9h-carbazole, 3,6-dibromo, carbazole, 3,6-dibromo, pubchem9951, 3,6-dibromo-carbazole, acmc-209o2m, 3,6,-dibromo-9h-carbazole, 3,6-dibromo-9-h-carbazole, ksc490o8t
PubChem CID 274874
IUPAC Name 3,6-dibromo-9H-carbazole
SMILES C1=CC2=C(C=C1Br)C3=C(N2)C=CC(=C3)Br

Specifications

Melting Point 204°C to 206°C
Quantity 25 g
Solubility Information Soluble in benzene,chloroform
Formula Weight 325
Percent Purity 99%
Chemical Name or Material 3,6-Dibromocarbazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only