missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromocarbazole, 97%

Catalog No. AC433970050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433970050 Supplier Thermo Scientific Chemicals Supplier No. 433970050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6825-20-3 | C12H7Br2N | 325 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6825-20-3
Molecular Formula C12H7Br2N
Molecular Weight (g/mol) 325
MDL Number MFCD00004961
InChI Key FIHILUSWISKVSR-UHFFFAOYSA-N
Synonym 3,6-dibromocarbazole, 9h-carbazole, 3,6-dibromo, carbazole, 3,6-dibromo, pubchem9951, 3,6-dibromo-carbazole, acmc-209o2m, 3,6,-dibromo-9h-carbazole, 3,6-dibromo-9-h-carbazole, ksc490o8t
PubChem CID 274874
IUPAC Name 3,6-dibromo-9H-carbazole
SMILES C1=CC2=C(C=C1Br)C3=C(N2)C=CC(=C3)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 212.0°C to 214.0°C
Color Tan
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 325
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 6-Dibromocarbazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only