accessibility menu, dialog, popup

UserName

CAS RN 6804-07-5

CH3OONNNONOH(E/Z)(E/Z)

CAS RN 6804-07-5

IUPAC Name: 4-hydroxy-3-({[(methoxycarbonyl)imino]amino}methylidene)-3,4-dihydroquinoxalin-1-ium-1-olate
Synonyms: carbadox
Molecular Weight (g/mol): 262.23
Molecular Formula: C11H10N4O4
InChi Key: BPMVRAQIQQEBLN-UHFFFAOYSA-N
SMILES: COC(=O)N=NC=C1C=[N+]([O-])C2=CC=CC=C2N1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results