accessibility menu, dialog, popup

UserName

NH2OHNO

CAS RN 68-41-7

IUPAC Name: 4-amino-1,2-oxazolidin-3-one
Synonyms: (+-)-4-amino-3-isoxazolidinone
Molecular Weight (g/mol): 102.09
Molecular Formula: C3H6N2O2
InChi Key: DYDCUQKUCUHJBH-UHFFFAOYNA-N
SMILES: NC1CONC1=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)