accessibility menu, dialog, popup

UserName

CAS RN 677-22-5

IUPAC Name: tert-butyl(chloro)magnesium
Synonyms: tert-butyl(chloro)magnesium
Molecular Weight (g/mol): 116.87
Molecular Formula: C4H9ClMg
InChi Key: ZDRJSYVHDMFHSC-UHFFFAOYSA-M
SMILES: CC(C)(C)[Mg]Cl
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)