accessibility menu, dialog, popup

UserName

CH3

CAS RN 673-32-5

IUPAC Name: (prop-1-yn-1-yl)benzene
Synonyms: benzene, 1-propynyl-
Molecular Weight (g/mol): 116.16
Molecular Formula: C9H8
InChi Key: GHUURDQYRGVEHX-UHFFFAOYSA-N
SMILES: CC#CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)