missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-propyne 97.0+%, TCI America™

Catalog No. P02195ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02195ML 5 mL
P021925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02195ML Supplier TCI America Supplier No. P02195ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 673-32-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00009272
InChI Key GHUURDQYRGVEHX-UHFFFAOYSA-N
Synonym 1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
PubChem CID 69601
IUPAC Name prop-1-ynylbenzene
SMILES CC#CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 181°C
Quantity 5 mL
Formula Weight 116.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-propyne