missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-propyne, 98%

Catalog No. AAA1427209
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1427209 10 g
AAA1427206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1427209 Supplier Thermo Scientific Chemicals Supplier No. A1427209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 673-32-5 | C9H8 | 116.163 g/mol

1-Phenyl-1-propyne is used as a intermediate for phrama industry and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1-propyne is used as a intermediate for phrama industry and chemical research.

Solubility
Sparingly soluble in water (0.26 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 673-32-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00009272
InChI Key GHUURDQYRGVEHX-UHFFFAOYSA-N
Synonym 1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
PubChem CID 69601
IUPAC Name prop-1-ynylbenzene
SMILES CC#CC1=CC=CC=C1

Specifications

Density 0.933
Boiling Point 183°C to 185°C
Flash Point 62°C (143°F)
Refractive Index 1.564
Quantity 10 g
Beilstein 1071474
Solubility Information Sparingly soluble in water (0.26g/L) (25°C).
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 1-Phenyl-1-propyne
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-607-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only