accessibility menu, dialog, popup

UserName

CAS RN 6708-37-8

IUPAC Name: 4-oxatricyclo[5.2.2.0²,⁶]undec-8-ene-3,5-dione
Synonyms: 4-oxatricyclo[5.2.2.0²,⁶]undec-8-ene-3,5-dione
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: YIHKILSPWGDWPR-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2C1C1CCC2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results