missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic Anhydride 98.0+%, TCI America™

Catalog No. B30385G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30385G Supplier TCI America Supplier No. B30385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6708-37-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
InChI Key YIHKILSPWGDWPR-UHFFFAOYSA-N
PubChem CID 220977
SMILES C1CC2C=CC1C3C2C(=O)OC3=O

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Chemical Name or Material Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic Anhydride