accessibility menu, dialog, popup

UserName

CAS RN 67004-64-2

CH3NH(S)HO

CAS RN 67004-64-2

IUPAC Name: (2S)-2-(2-hydroxyethyl)-1-methylpyrrolidin-1-ium
Synonyms: (2S)-2-(2-hydroxyethyl)-1-methylpyrrolidin-1-ium
Molecular Weight (g/mol): 130.21
Molecular Formula: C7H16NO
InChi Key: FYVMBPXFPFAECB-ZETCQYMHSA-O
SMILES: C[NH+]1CCC[C@H]1CCO
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)