missing translation for 'onlineSavingsMsg'
Learn More

Cholecalciferol (Vitamin D3) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019154
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019154 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019154 Supplier Merck Emd Millipore Supplier No. PHR1237500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Cholecalciferol (vitamin D3) is a fat-soluble vitamin that manifests skeletal and extra-skeletal effects. The skeletal effects augment the bone health whereas the extra-skeletal effects concern the immune system, the cognition, the cardiovascular system, etc. Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 67-97-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C27H44 O
MDL Number MFCD00078131
Melting Point 83°C to 86°C
Synonym Cholecalciferol; (+)-Vitamin D3; 7-Dehydrocholesterol activated; Activated 7-dehydrocholesterol; Calciol
Type Certified Reference Material
Quantity 500 mg
Formula Weight 384.64
Grade Certified Reference Material
Show More Show Less