accessibility menu, dialog, popup

UserName

H3CH3CO(R)(S)(S)(S)FO(S)H3C(R)F(S)OH(S)H3C(S)OOHO

CAS RN 67-73-2

IUPAC Name: (1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Synonyms: fluocinolone acetonide fluocinolone 16,17-acetonide Synalar-HP medicis brand 2 of fluocinolone acetonide synemol cortiplastol fluocinoloni acetonidum allergan brand of fluocinolone acetonide percutina inkeysa brand of fluocinolone acetonide
Molecular Weight (g/mol): 452.5
Molecular Formula: C24H30F2O6
InChi Key: FEBLZLNTKCEFIT-VSXGLTOVSA-N
SMILES: CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results