accessibility menu, dialog, popup

UserName

CAS RN 67-47-0

IUPAC Name: 5-(hydroxymethyl)furan-2-carbaldehyde
Synonyms: hydroxymethylfurfural 5-(hydroxymethyl)-2-furfural HMF hydroxymethylfurfuraldehyde hydroxymethylfurfuralaldehyde hydroxymethylfurfurole 5-hydroxymethyl-2-furfural 5-(hyddroxymethyl)furfurole 2-hydroxymethyl-5-furfural 5-(hydroxymethyl)furfurole
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: NOEGNKMFWQHSLB-UHFFFAOYSA-N
SMILES: OCC1=CC=C(O1)C=O
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)