missing translation for 'onlineSavingsMsg'
Learn More

5-(Hydroxymethyl)furfural, ≥98% (GC/HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015133
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015133 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015133 Supplier Merck Emd Millipore Supplier No. 53407100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-(Hydroxymethyl)furfural is an organic compound, widely spread in food products and is formed in sugar containing food when exposed to heat. It is widely used as an indicator of quality in food products. It is also employed to indicate the adulteration of food products with acid converted invert syrups. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 67-47-0
Density 1.243 g/mL (at 25°C)
Flash Point 79°C
Physical Form Neat
Linear Formula C6H6O3
MDL Number MFCD00003234
Melting Point 28°C to 34°C; 28°C to 34°C
Boiling Point 114°C to 116°C (1 mmHg)
Synonym 5-Hydroxymethyl-2-furaldehyde; 5-(Hydroxymethyl)furfural; 5-Hydroxymethyl-2-furancarboxaldehyde; HMF
Type Analytical Standard
Quantity 100 mg
Formula Weight 126.11
Percent Purity ≥98% (GC/HPLC)
Grade Analytical Standard
Show More Show Less