accessibility menu, dialog, popup

UserName

CAS RN 66612-29-1

IUPAC Name: 6-[(4-aminobutyl)(ethyl)amino]-1,2,3,4-tetrahydrophthalazine-1,4-dione
Synonyms: 6-[(4-aminobutyl)(ethyl)amino]-2,3-dihydrophthalazine-1,4-dione
Molecular Weight (g/mol): 276.34
Molecular Formula: C14H20N4O2
InChi Key: LEOJISUPFSWNMA-UHFFFAOYSA-N
SMILES: CCN(CCCCN)C1=CC=C2C(=O)NNC(=O)C2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)