accessibility menu, dialog, popup

UserName

H3CCH3H3COONH(S)OH

CAS RN 66605-57-0

IUPAC Name: tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol): 251.33
Molecular Formula: C14H21NO3
InChi Key: LDKDMDVMMCXTMO-LBPRGKRZSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)