missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylalaninol, 99%

Catalog No. AAB2501914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2501914 25 g
AAB2501903 1 g
AAB2501906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2501914 Supplier Thermo Scientific Chemicals Supplier No. B2501914

CAS: 66605-57-0 | C14H21NO3 | 251.33 g/mol

N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 66605-57-0
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.33
MDL Number MFCD00076976
InChI Key LDKDMDVMMCXTMO-LBPRGKRZSA-N
Synonym n-boc-l-phenylalaninol, boc-l-phenylalaninol, boc-phenylalaninol, s-2-boc-amino-3-phenyl-1-propanol, n-tert-butoxycarbonyl-l-phenylalaninol, tert-butyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, s-n-tert-butoxycarbonyl-phenylalaninol, s---2-tert-butoxycarbonylamino-3-phenyl-1-propanol
PubChem CID 2733675
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 95°C to 98°C
Beilstein 3652004
Solubility Information Insoluble in water.
Quantity 25 g
Optical Rotation −27.5° ± 2.5° (c=1 in Methanol)
Formula Weight 251.33
Percent Purity 99%
Chemical Name or Material N-Boc-L-phenylalaninol
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only