missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-phenylalaninol 97.0+%, TCI America™

Catalog No. B26045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26045G Supplier TCI America Supplier No. B26045G

Chemical Identifiers

CAS 66605-57-0
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.33
MDL Number MFCD00076976
InChI Key LDKDMDVMMCXTMO-LBPRGKRZSA-N
Synonym n-boc-l-phenylalaninol, boc-l-phenylalaninol, boc-phenylalaninol, s-2-boc-amino-3-phenyl-1-propanol, n-tert-butoxycarbonyl-l-phenylalaninol, tert-butyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, s-n-tert-butoxycarbonyl-phenylalaninol, s---2-tert-butoxycarbonylamino-3-phenyl-1-propanol
PubChem CID 2733675
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 96°C
Color White
Quantity 5 g
Formula Weight 251.33
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-phenylalaninol